siRNA Details
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181639.3 | NP_853670.2 | 2 | 309 | ||
GenBank mRNA | AY273196.1 | - | 1 | 231 | ||
BC078678.1 | - | 2 | 309 | 7132846 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181639.3 | NP_853670.2 | 2 | 309 | ||
GenBank mRNA | AY273196.1 | - | 1 | 231 | ||
BC078678.1 | - | 2 | 309 | 7132846 |