siRNA Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182672.2 | NP_872613.1 | 3 | 381 | ||
| GenBank mRNA | AY305858.1 | - | 3 | 382 | ||
| BC086378.1 | - | 3 | 348 | 7302014 | ||
| FQ215409.1 | - | 3 | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbr4| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182672.2 | NP_872613.1 | 3 | 381 | ||
| GenBank mRNA | AY305858.1 | - | 3 | 382 | ||
| BC086378.1 | - | 3 | 348 | 7302014 | ||
| FQ215409.1 | - | 3 | 402 | |||