siRNA Details
| Gene Symbol: Bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182674.3 | NP_872615.3 | 7 | 1105 | ||
| GenBank mRNA | AY257675.4 | - | 7 | 1088 | ||
| BC079030.1 | - | 7 | 1094 | 7114086 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bysl| Gene Symbol: Bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182674.3 | NP_872615.3 | 7 | 1105 | ||
| GenBank mRNA | AY257675.4 | - | 7 | 1088 | ||
| BC079030.1 | - | 7 | 1094 | 7114086 | ||