siRNA Details
| Gene Symbol: Brca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031542.2 | NP_113730.2 | 11 | 2418 | ||
| GenBank mRNA | U89653.1 | - | 11 | 2417 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brca2| Gene Symbol: Brca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031542.2 | NP_113730.2 | 11 | 2418 | ||
| GenBank mRNA | U89653.1 | - | 11 | 2417 | ||