siRNA Details
| Gene Symbol: Ptprf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019249.1 | NP_062122.1 | 4 | 567 | ||
| XM_006238677.2 | XP_006238739.1 | 6 | 1583 | |||
| XM_006238678.2 | XP_006238740.1 | 6 | 1583 | |||
| XM_006238680.2 | XP_006238742.1 | 6 | 1583 | |||
| XM_006238681.2 | XP_006238743.1 | 6 | 1583 | |||
| GenBank mRNA | L11586.1 | - | 4 | 567 | ||

Ptprf
Genomic Map