siRNA Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133543.3 | NP_598227.3 | 722 | |||
| GenBank mRNA | BC088090.1 | - | 722 | 7368214 | ||
| BC098650.1 | - | 784 | 7371059 | |||
| FQ209576.1 | - | 706 | ||||
| FQ209710.1 | - | 740 | ||||
| U18762.1 | - | 931 | ||||
| U33501.1 | - | 703 | ||||

Rdh7
Genomic Map