siRNA Details
| Gene Symbol: Crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011710.2 | NP_001011710.1 | 10 | 1005 | ||
| XM_006244643.2 | XP_006244705.1 | 9 | 988 | |||
| GenBank mRNA | AB009662.1 | - | 9 | 868 | ||
| BC078799.1 | - | 10 | 954 | 7112769 | ||

Crisp2
Genomic Map