siRNA Details
| Gene Symbol: Fopnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108261.1 | NP_001101731.1 | 2 | 137 | ||
| XM_006245848.2 | XP_006245910.1 | 2 | 166 | |||
| GenBank mRNA | BC083733.1 | - | 2 | 111 | ||
| BC158662.1 | - | 2 | 103 | 8369763 | ||
| FQ215640.1 | - | 2 | 103 | |||
| FQ228774.1 | - | 2 | 111 | |||
| FQ228837.1 | - | 2 | 93 | |||

Fopnl
Genomic Map