siRNA Details
| Gene Symbol: Parn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270415.1 | NP_001257344.1 | 254 | |||
| GenBank mRNA | HQ123458.1 | - | 254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parn| Gene Symbol: Parn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270415.1 | NP_001257344.1 | 254 | |||
| GenBank mRNA | HQ123458.1 | - | 254 | |||