siRNA Details
| Gene Symbol: Zc3h7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108262.1 | NP_001101732.1 | 5 | 521 | ||
| XM_008767489.1 | XP_008765711.1 | 5 | 1050 | |||
| XM_008767490.1 | XP_008765712.1 | 6 | 1244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h7a| Gene Symbol: Zc3h7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108262.1 | NP_001101732.1 | 5 | 521 | ||
| XM_008767489.1 | XP_008765711.1 | 5 | 1050 | |||
| XM_008767490.1 | XP_008765712.1 | 6 | 1244 | |||