siRNA Details
| Gene Symbol: DGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012079.5 | NP_036211.2 | 14, 15 | 1450 | ||
| XM_011517356.2 | XP_011515658.1 | 14, 15 | 1171 | |||
| GenBank mRNA | AB057815.1 | - | 14, 15 | 1193 | ||
| AF059202.1 | - | 14, 15 | 1411 | |||
| BC006263.2 | - | 13, 14 | 903 | |||
| BC015762.1 | - | 14, 15 | 1415 | 4863905 | ||
| BC023565.1 | - | 14, 15 | 1420 | 4130484 | ||
| BC150649.1 | - | 14, 15 | 1396 | 9057020 | ||

DGAT1
Genomic Map