siRNA Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007800.1 | NP_001007801.1 | 9, 10 | 802 | ||
| XM_006245803.2 | XP_006245865.1 | 9, 10 | 871 | |||
| XM_006245804.2 | XP_006245866.1 | 8, 9 | 631 | |||
| GenBank mRNA | BC085931.1 | - | 9, 10 | 813 | 7111787 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028720, NM_001079814, CF174076, AK208357, AK150349, and others.

Glyr1
Genomic Map