siRNA Details
| Gene Symbol: Dnaja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038595.1 | NP_001033684.1 | 5 | 687 | ||
| NM_001038596.1 | NP_001033685.1 | 5 | 687 | |||
| GenBank mRNA | AF516338.1 | - | 5 | 687 | ||
| AY077460.1 | - | 5 | 687 | |||

Dnaja3
Genomic Map