siRNA Details
| Gene Symbol: Dnaja3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038595.1 | NP_001033684.1 | 9 | 1144 | ||
| NM_001038596.1 | NP_001033685.1 | 9 | 1144 | |||
| XM_006245817.2 | XP_006245879.1 | 9 | 1184 | |||
| GenBank mRNA | AF516338.1 | - | 9 | 1144 | ||
| AY077460.1 | - | 9 | 1144 | |||
| AY264842.1 | - | 8 | 991 | |||

Dnaja3
Genomic Map