siRNA Details
| Gene Symbol: Tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108268.1 | NP_001101738.1 | 2, 3 | 674 | ||
| XM_006245996.2 | XP_006246058.1 | 3, 4 | 585 | |||
| XM_006245997.2 | XP_006246059.1 | 3, 4 | 564 | |||
| GenBank mRNA | BC167035.1 | - | 1, 2 | 802 | ||

Tsr3
Genomic Map