siRNA Details
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034936.1 | NP_001030108.1 | 3 | 365 | ||
| XM_006246008.2 | XP_006246070.1 | 4 | 487 | |||
| XM_006246009.2 | XP_006246071.1 | 4 | 487 | |||
| XM_006246010.2 | XP_006246072.1 | 3 | 447 | |||
| XM_006246011.2 | XP_006246073.1 | 4 | 299 | |||
| XM_006246013.2 | XP_006246075.1 | 2 | 1929 | |||
| XM_006246014.2 | XP_006246076.1 | 2 | 1830 | |||
| XM_006246015.2 | XP_006246077.1 | 2 | 2131 | |||
| XM_006246016.2 | XP_006246078.1 | 4 | 499 | |||
| GenBank mRNA | BC099803.1 | - | 3 | 365 | 7937864 | |

Nprl3
Genomic Map