siRNA Details
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034936.1 | NP_001030108.1 | 4 | 456 | ||
| XM_006246008.2 | XP_006246070.1 | 5 | 578 | |||
| XM_006246009.2 | XP_006246071.1 | 5 | 578 | |||
| XM_006246010.2 | XP_006246072.1 | 4 | 538 | |||
| XM_006246011.2 | XP_006246073.1 | 5 | 390 | |||
| XM_006246013.2 | XP_006246075.1 | 3 | 2020 | |||
| XM_006246014.2 | XP_006246076.1 | 3 | 1921 | |||
| XM_006246015.2 | XP_006246077.1 | 3 | 2222 | |||
| XM_006246016.2 | XP_006246078.1 | 5 | 590 | |||
| XM_006246017.2 | XP_006246079.1 | 2 | 112 | |||
| GenBank mRNA | BC099803.1 | - | 4 | 456 | 7937864 | |

Nprl3
Genomic Map