siRNA Details
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034936.1 | NP_001030108.1 | 2 | 235 | ||
| XM_006246008.2 | XP_006246070.1 | 2 | 318 | |||
| XM_006246009.2 | XP_006246071.1 | 2 | 318 | |||
| XM_006246010.2 | XP_006246072.1 | 2 | 317 | |||
| XM_006246011.2 | XP_006246073.1 | 2 | 130 | |||
| XM_006246016.2 | XP_006246078.1 | 2 | 330 | |||
| XM_006246017.2 | XP_006246079.1 | 1 | 21 | |||
| GenBank mRNA | BC099803.1 | - | 2 | 235 | 7937864 | |

Nprl3
Genomic Map