siRNA Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014117.1 | NP_001014139.1 | 5 | 515 | ||
| XM_006246140.1 | XP_006246202.1 | 5 | 545 | |||
| GenBank mRNA | BC089210.1 | - | 5 | 515 | 7302690 | |
| FQ233702.1 | - | 5 | 518 | |||
| FQ233841.1 | - | 5 | 518 | |||

Ublcp1
Genomic Map