siRNA Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002819.2 | NP_001002819.2 | 4 | 332 | ||
| GenBank mRNA | BC098630.1 | - | 4 | 332 | 7105780 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfpt2| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002819.2 | NP_001002819.2 | 4 | 332 | ||
| GenBank mRNA | BC098630.1 | - | 4 | 332 | 7105780 | |