siRNA Details
| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100727.1 | NP_001094197.1 | 10 | 1223 | ||
| XM_006246298.2 | XP_006246360.1 | 10 | 1280 | |||
| GenBank mRNA | BC099227.1 | - | 4 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rufy1| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100727.1 | NP_001094197.1 | 10 | 1223 | ||
| XM_006246298.2 | XP_006246360.1 | 10 | 1280 | |||
| GenBank mRNA | BC099227.1 | - | 4 | 256 | ||