siRNA Details
| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271026.1 | NP_001257955.1 | 2 | 63 | ||
| GenBank mRNA | AY387053.1 | - | 509 | |||
| BC089894.1 | - | 2 | 63 | |||
| BC158679.1 | - | 2 | 63 | |||
| FQ227271.1 | - | 2 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc10| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271026.1 | NP_001257955.1 | 2 | 63 | ||
| GenBank mRNA | AY387053.1 | - | 509 | |||
| BC089894.1 | - | 2 | 63 | |||
| BC158679.1 | - | 2 | 63 | |||
| FQ227271.1 | - | 2 | 84 | |||