siRNA Details
| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271026.1 | NP_001257955.1 | 2 | 119 | ||
| GenBank mRNA | AY387053.1 | - | 565 | |||
| BC089894.1 | - | 2 | 119 | |||
| BC158679.1 | - | 2 | 119 | |||
| FQ227271.1 | - | 2 | 140 | |||

Zcchc10
Genomic Map