siRNA Details
| Gene Symbol: Rnf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164264.1 | NP_001157736.1 | 3 | 670 | ||
| GenBank mRNA | DY320214.1 | - | 3 | 318 | ||
| DY569693.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf187| Gene Symbol: Rnf187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164264.1 | NP_001157736.1 | 3 | 670 | ||
| GenBank mRNA | DY320214.1 | - | 3 | 318 | ||
| DY569693.1 | - | 1 | 47 | |||