siRNA Details
| Gene Symbol: Plscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012139.2 | NP_001012139.1 | 4 | 486 | ||
| XM_008767838.1 | XP_008766060.1 | 3 | 837 | |||
| XM_008767839.1 | XP_008766061.1 | 3 | 1821 | |||
| GenBank mRNA | AY548831.1 | - | 3 | 363 | ||
| BC098055.1 | - | 4 | 486 | 7457695 | ||

Plscr3
Genomic Map