siRNA Details
| Gene Symbol: Bcl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108279.1 | NP_001101749.1 | 2 | 302 | ||
| XM_008767841.1 | XP_008766063.1 | 3 | 397 | |||
| XM_008767842.1 | XP_008766064.1 | 3 | 687 | |||
| XM_008767843.1 | XP_008766065.1 | 3 | 387 | |||
| GenBank mRNA | FQ223808.1 | - | 3 | 358 | ||
| FQ224714.1 | - | 3 | 358 | |||

Bcl6b
Genomic Map