siRNA Details
| Gene Symbol: Inca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108280.1 | NP_001101750.1 | 6 | 632 | ||
| XM_006246751.1 | XP_006246813.1 | 5 | 662 | |||
| XM_006246752.2 | XP_006246814.1 | 5 | 746 | |||
| XM_006246754.2 | XP_006246816.1 | 5 | 453 | |||
| XM_008767844.1 | XP_008766066.1 | 4 | 816 | |||

Inca1
Genomic Map