siRNA Details
| Gene Symbol: RGD1304728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246849.2 | XP_006246911.1 | 3 | 998 | ||
| XM_006246853.2 | XP_006246915.1 | 3 | 998 | |||
| XM_008767904.1 | XP_008766126.1 | 3 | 847 | |||
| XM_008767905.1 | XP_008766127.1 | 3 | 763 | |||
| XM_008767906.1 | XP_008766128.1 | 3 | 971 | |||
| XM_008767907.1 | XP_008766129.1 | 3 | 998 | |||
| XM_340837.6 | XP_340838.3 | 3 | 998 | |||

RGD1304728
Genomic Map