siRNA Details
| Gene Symbol: Rnmtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108282.1 | NP_001101752.1 | 1 | 235 | ||
| GenBank mRNA | FQ232324.1 | - | 1 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnmtl1| Gene Symbol: Rnmtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108282.1 | NP_001101752.1 | 1 | 235 | ||
| GenBank mRNA | FQ232324.1 | - | 1 | 233 | ||