siRNA Details
| Gene Symbol: Tusc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039163.1 | NP_001034252.1 | 1 | 628 | ||
| GenBank mRNA | AB218813.1 | - | 1 | 628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tusc5| Gene Symbol: Tusc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039163.1 | NP_001034252.1 | 1 | 628 | ||
| GenBank mRNA | AB218813.1 | - | 1 | 628 | ||