siRNA Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012141.1 | NP_001012141.1 | 5, 6 | 453 | ||
| XM_006247041.2 | XP_006247103.1 | 4, 5 | 416 | |||
| XM_006247042.2 | XP_006247104.1 | 4, 5 | 362 | |||
| GenBank mRNA | BC079084.1 | - | 5, 6 | 453 | 7123007 | |
| EF121983.1 | - | 4, 5 | 292 | |||
| EF121984.1 | - | 4, 5 | 292 | |||
| EF121985.1 | - | 4, 5 | 292 | |||

Tada2a
Genomic Map