siRNA Details
| Gene Symbol: Dhrs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014119.2 | NP_001014141.2 | 4 | 643 | ||
| XM_006247045.2 | XP_006247107.1 | 4 | 764 | |||
| GenBank mRNA | BC088444.1 | - | 4 | 637 | 7126427 | |
| DY311021.1 | - | 4 | 643 | |||

Dhrs11
Genomic Map