siRNA Details
| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108287.1 | NP_001101757.1 | 5, 6 | 725 | ||
| XM_008768073.1 | XP_008766295.1 | 5, 6 | 882 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr11| Gene Symbol: Prr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108287.1 | NP_001101757.1 | 5, 6 | 725 | ||
| XM_008768073.1 | XP_008766295.1 | 5, 6 | 882 | |||