siRNA Details
| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199402.1 | NP_955434.1 | 2 | 169 | ||
| XM_006247200.2 | XP_006247262.1 | 2 | 195 | |||
| GenBank mRNA | AY438568.1 | - | 2 | 169 | ||
| BC128718.1 | - | 2 | 129 | |||
| BC158571.1 | - | 2 | 128 | |||

Spata20
Genomic Map