siRNA Details
| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199402.1 | NP_955434.1 | 15 | 2250 | ||
| XM_006247200.2 | XP_006247262.1 | 15 | 2276 | |||
| GenBank mRNA | AY438568.1 | - | 15 | 2250 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata20| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199402.1 | NP_955434.1 | 15 | 2250 | ||
| XM_006247200.2 | XP_006247262.1 | 15 | 2276 | |||
| GenBank mRNA | AY438568.1 | - | 15 | 2250 | ||