siRNA Details
| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750907.2 | XP_003750955.1 | 2 | 744 | ||
| XM_006247215.2 | XP_006247277.1 | 2 | 744 | |||
| XM_008768184.1 | XP_008766406.1 | 3 | 702 | |||
| GenBank mRNA | AB727941.1 | - | 2 | 322 | ||
| AB727942.1 | - | 2 | 322 | |||

Itga3
Genomic Map