siRNA Details
| Gene Symbol: Copz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108294.1 | NP_001101764.1 | 4 | 299 | ||
| GenBank mRNA | FQ225734.1 | - | 4 | 303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copz2| Gene Symbol: Copz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108294.1 | NP_001101764.1 | 4 | 299 | ||
| GenBank mRNA | FQ225734.1 | - | 4 | 303 | ||