siRNA Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012142.1 | NP_001012142.1 | 2 | 132 | ||
| XM_006247342.2 | XP_006247404.1 | 2 | 361 | |||
| XM_008768079.1 | XP_008766301.1 | 2 | 1645 | |||
| GenBank mRNA | BC078939.1 | - | 2 | 132 | 7108372 | |

Scrn2
Genomic Map