siRNA Details
| Gene Symbol: Cwc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108295.1 | NP_001101765.1 | 7 | 854 | ||
| GenBank mRNA | BC168194.1 | - | 5 | 562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cwc25| Gene Symbol: Cwc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108295.1 | NP_001101765.1 | 7 | 854 | ||
| GenBank mRNA | BC168194.1 | - | 5 | 562 | ||