siRNA Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138521.1 | NP_612530.1 | 5 | 709 | ||
| XM_006247344.2 | XP_006247406.1 | 2 | 1207 | |||
| GenBank mRNA | AF281661.1 | - | 1 | 34 | ||
| AY601872.1 | - | 5 | 709 | |||
| BC078954.1 | - | 5 | 639 | 7110316 | ||

Ppp1r1b
Genomic Map