siRNA Details
| Gene Symbol: LOC102552549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247244.2 | XP_006247306.1 | 4 | 658 | ||
| Gene Symbol: Mien1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108296.1 | NP_001101766.1 | 4 | 345 | ||
| GenBank mRNA | FQ214212.1 | - | 342 | |||

LOC102552549
Genomic Map