siRNA Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108297.1 | NP_001101767.1 | 9 | 869 | ||
| XM_006247347.1 | XP_006247409.1 | 9 | 904 | |||
| GenBank mRNA | BC112385.1 | - | 8 | 1739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdma| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108297.1 | NP_001101767.1 | 9 | 869 | ||
| XM_006247347.1 | XP_006247409.1 | 9 | 904 | |||
| GenBank mRNA | BC112385.1 | - | 8 | 1739 | ||