siRNA Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191825.1 | NP_001178754.1 | 5 | 1156 | ||
| XM_006247353.2 | XP_006247415.1 | 6 | 1382 | |||
| XM_008768080.1 | XP_008766302.1 | 6 | 1383 | |||
| XM_008768081.1 | XP_008766303.1 | 6 | 1421 | |||
| XM_008768082.1 | XP_008766304.1 | 6 | 3066 | |||

Wipf2
Genomic Map