siRNA Details
| Gene Symbol: Cdc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108298.1 | NP_001101768.1 | 7, 8 | 1265 | ||
| XM_006247354.2 | XP_006247416.1 | 7, 8 | 1267 | |||
| XM_006247355.2 | XP_006247417.1 | 7, 8 | 1265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc6| Gene Symbol: Cdc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108298.1 | NP_001101768.1 | 7, 8 | 1265 | ||
| XM_006247354.2 | XP_006247416.1 | 7, 8 | 1267 | |||
| XM_006247355.2 | XP_006247417.1 | 7, 8 | 1265 | |||