siRNA Details
| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008760.1 | NP_001008760.1 | 1 | 368 | ||
| XM_006247356.2 | XP_006247418.1 | 1 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt28| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008760.1 | NP_001008760.1 | 1 | 368 | ||
| XM_006247356.2 | XP_006247418.1 | 1 | 599 | |||