siRNA Details
| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008761.1 | NP_001008761.1 | 7, 8 | 1281 | ||
| XM_006247357.2 | XP_006247419.1 | 7, 8 | 2293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt12| Gene Symbol: Krt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008761.1 | NP_001008761.1 | 7, 8 | 1281 | ||
| XM_006247357.2 | XP_006247419.1 | 7, 8 | 2293 | |||