siRNA Details
| Gene Symbol: Krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY464140.1 | - | 4 | 883 | ||
| BC088424.1 | - | 4 | 879 | |||
| BC126075.1 | - | 4 | 874 | 7442082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt19| Gene Symbol: Krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY464140.1 | - | 4 | 883 | ||
| BC088424.1 | - | 4 | 879 | |||
| BC126075.1 | - | 4 | 874 | 7442082 | ||