siRNA Details
| Gene Symbol: Slc25a39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024792.1 | NP_001019963.1 | 7 | 539 | ||
| XM_006247374.2 | XP_006247436.1 | 7 | 877 | |||
| XM_006247376.2 | XP_006247438.1 | 7 | 562 | |||
| GenBank mRNA | BC097349.1 | - | 7 | 539 | 7460172 | |
| FQ228904.1 | - | 7 | 529 | |||

Slc25a39
Genomic Map