siRNA Details
| Gene Symbol: Slc25a39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024792.1 | NP_001019963.1 | 12 | 1127 | ||
| XM_006247374.2 | XP_006247436.1 | 12 | 1465 | |||
| XM_006247376.2 | XP_006247438.1 | 12 | 1150 | |||
| GenBank mRNA | BC081862.1 | - | 12 | 1044 | ||
| BC097349.1 | - | 12 | 1127 | 7460172 | ||
| FQ228904.1 | - | 12 | 1119 | |||

Slc25a39
Genomic Map