siRNA Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100728.1 | NP_001094198.1 | 2 | 267 | ||
| GenBank mRNA | BC091255.1 | - | 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc43| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100728.1 | NP_001094198.1 | 2 | 267 | ||
| GenBank mRNA | BC091255.1 | - | 2 | 241 | ||